missing translation for 'onlineSavingsMsg'
Learn More

2-Methoxy-5-nitrophenol, 98%, Thermo Scientific™

Product Code. 10113404 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 10113404

Brand: Thermo Scientific Acros 397540050

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 636-93-1
Molecular Formula C7H7NO4
Molecular Weight (g/mol) 169.14
MDL Number MFCD00015561
InChI Key KXKCTSZYNCDFFG-UHFFFAOYSA-N
Synonym 5-nitroguaiacol, phenol, 2-methoxy-5-nitro, 3-hydroxy-4-methoxynitrobenzene, 2-methoxy-5-nitro-phenol, unii-e5t11i2jub, e5t11i2jub, 2-hydroxy-4-nitroanisole, acmc-1b3lw, 2-methoxy-5-nitro phenol, 2-methyloxy-5-nitrophenol
PubChem CID 69471
ChEBI CHEBI:81049
IUPAC Name 2-methoxy-5-nitrophenol
SMILES COC1=C(C=C(C=C1)[N+](=O)[O-])O

Specifications

Melting Point 103°C to 107°C
Boiling Point 110°C to 112°C (0.8 mmHg)
Assay Percent Range 97.5% min. (GC)
Packaging Glass Bottle
Linear Formula CH3OC6H3(NO2)OH
Quantity 5 g
Solubility Information Solubility in water: slightly soluble
Formula Weight 169.14
Percent Purity 98%
Chemical Name or Material 2-Methoxy-5-nitrophenol
compliance-icons
Product Identifier
  • 2-Methoxy-5-nitrophenol
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P264-Wash thoroughly after handling.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P363-Wash contaminated clothing before reuse.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.