missing translation for 'onlineSavingsMsg'
Learn More

2-Methoxy-5-nitrophenol, 98%, Thermo Scientific™

Product Code. 10254834 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10254834 25 g 25g
10113404 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10254834 Supplier Thermo Scientific Acros Supplier No. 397540250
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 636-93-1
Molecular Formula C7H7NO4
Molecular Weight (g/mol) 169.14
MDL Number MFCD00015561
InChI Key KXKCTSZYNCDFFG-UHFFFAOYSA-N
Synonym 5-nitroguaiacol, phenol, 2-methoxy-5-nitro, 3-hydroxy-4-methoxynitrobenzene, 2-methoxy-5-nitro-phenol, unii-e5t11i2jub, e5t11i2jub, 2-hydroxy-4-nitroanisole, acmc-1b3lw, 2-methoxy-5-nitro phenol, 2-methyloxy-5-nitrophenol
PubChem CID 69471
ChEBI CHEBI:81049
IUPAC Name 2-methoxy-5-nitrophenol
SMILES COC1=C(C=C(C=C1)[N+](=O)[O-])O

Specifications

Melting Point 103°C to 107°C
Boiling Point 110°C to 112°C (0.8 mmHg)
Assay Percent Range 97.5% min. (GC)
Packaging Glass Bottle
Linear Formula CH3OC6H3(NO2)OH
Quantity 25 g
Solubility Information Solubility in water: slightly soluble
Formula Weight 169.14
Percent Purity 98%
Chemical Name or Material 2-Methoxy-5-nitrophenol
compliance-icons compliance-icons
Product Identifier
  • 2-Methoxy-5-nitrophenol
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.