missing translation for 'onlineSavingsMsg'
Learn More

(2-Methoxy-5-nitrophenyl)acetic acid, 97%

Product Code. 15497317
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15497317 250 mg 250mg
15477317 1 g 1g
15487317 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15497317 Supplier Thermo Scientific Alfa Aesar Supplier No. H34386.MD
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 51073-04-2 | C9H9NO5 | 211.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 51073-04-2
Molecular Formula C9H9NO5
Molecular Weight (g/mol) 211.17
MDL Number MFCD02664832
InChI Key TZQZNDOEOCOJFS-UHFFFAOYSA-N
Synonym 2-2-methoxy-5-nitrophenyl acetic acid, 2-methoxy-5-nitrophenyl acetic acid, 2-methoxy-5-nitrophenylacetic acid, benzeneacetic acid, 2-methoxy-5-nitro, 2-methoxy-5-nitro-phenyl-acetic acid
PubChem CID 3519298
IUPAC Name 2-(2-methoxy-5-nitrophenyl)acetic acid
SMILES COC1=C(CC(O)=O)C=C(C=C1)[N+]([O-])=O

Specifications

Melting Point 153°C to 155°C
Quantity 250 mg
Formula Weight 211.17
Percent Purity 97%
Chemical Name or Material (2-Methoxy-5-nitrophenyl)acetic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.