missing translation for 'onlineSavingsMsg'
Learn More

2-Methoxybenzeneboronic acid, 97%

Product Code. 11440167
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Quantity unitSize
11440167 1 g 1g
11454517 5 g 5g
11464517 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Artikelnummer. 11440167 Lieferant Thermo Scientific Alfa Aesar Lieferanten-Nr. B21071.03
 Restricted Item
Estimated Shipment: 24-07-2026
Zum Warenkorb hinzufügen
Zum Warenkorb hinzufügen
Request Bulk or Custom Format
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

CAS: 5720-06-9 | C7H9BO3 | 151.96 g/mol

2-Methoxylphenylboronic acid is a phenylboronic acid used to investigate boron function in plants. It is used in Suzuki reaction, organic reaction that is classified as a coupling reaction where the coupling partners are a boronic acid with a halide catalyzed by a palladium(0) complex.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Methoxylphenylboronic acid is a phenylboronic acid used to investigate boron function in plants. It is used in Suzuki reaction, organic reaction that is classified as a coupling reaction where the coupling partners are a boronic acid with a halide catalyzed by a palladium(0) complex.

Solubility
Insoluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemische Identifikatoren

CAS 5720-06-9
Molecular Formula C7H9BO3
Molecular Weight (g/mol) 151.96
MDL Number MFCD00236047
InChI Key ROEQGIFOWRQYHD-UHFFFAOYSA-N
Synonym 2-methoxybenzeneboronic acid, 2-methoxyphenyl boronic acid, o-methoxyphenylboronic acid, 2-methoxyphenyl boranediol, 2-boronoanisole, 2-methoxypenylboronic acid, o-methoxyphenyl boronic acid, 2-methoxy phenylboronic acid, 2-methoxybenzene boronic acid
PubChem CID 2733958
IUPAC Name (2-methoxyphenyl)boronic acid
SMILES COC1=CC=CC=C1B(O)O

Spezifikation

Melting Point 102°C to 106°C
Quantity 1 g
Beilstein 3030981
Solubility Information Insoluble in water.
Formula Weight 151.96
Percent Purity 97%
Chemical Name or Material 2-Methoxybenzeneboronic acid

RUO – Research Use Only

Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.