missing translation for 'onlineSavingsMsg'
Learn More

2-Methoxybenzenethiol, 97%, Thermo Scientific™

Product Code. 10647114 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 mL
Packaging:
Glass bottle
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10647114 1 mL Glass bottle 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10647114 Supplier Thermo Scientific Acros Supplier No. 179980010
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7217-59-6
Molecular Formula C7H8OS
Molecular Weight (g/mol) 140.2
MDL Number MFCD00004834
InChI Key DSCJETUEDFKYGN-UHFFFAOYSA-N
Synonym 2-methoxythiophenol, thioguaiacol, benzenethiol, 2-methoxy, o-methoxythiophenol, o-methoxybenzenethiol, 2-methoxy thiophenol, 2-mercaptoanisole, 2-methoxybenzene-1-thiol, 2-methoxy-benzenethiol, benzenethiol, o-methoxy
PubChem CID 23642
IUPAC Name 2-methoxybenzenethiol
SMILES COC1=CC=CC=C1S

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Yellow
Density 1.1400g/mL
Boiling Point 225.0°C to 228.0°C
Flash Point 95°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula CH3OC6H4SH
Refractive Index 1.5905 to 1.5925
Quantity 1 mL
Specific Gravity 1.14
Solubility Information Solubility in water: insoluble in water
Formula Weight 140.2
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 2-Methoxybenzenethiol, 97%
Afficher plus Afficher moins
compliance-icons
Identifiant produit
  • 2-Methoxybenzenethiol
Mention d'avertissement
  • Warning
Catégorie de danger
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Mention de danger
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Conseils de prudence
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.