missing translation for 'onlineSavingsMsg'
Learn More

2-Methoxybenzoic acid, MedChemExpress

Product Code. 16422569
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16422569 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16422569 Supplier Medchem Express Supplier No. HYN1393
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

MedChemExpress 2-Methoxybenzoic acid (NSC 3778) is used as an internal standard of salicylic acid and its putative biosynthetic precursors in cucumber leaves. Another known use is in the synthesis of Benextramine.

2-Methoxybenzoic acid (NSC 3778) is used as an internal standard of salicylic acid and its putative biosynthetic precursors in cucumber leaves. Another known use is in the synthesis of Benextramine.

Specifications

Chemical Name or Material 2-Methoxybenzoic acid
CAS 579-75-9
Color White
Purity Grade Notes Research
Molecular Formula C8H8O3
Quantity 1 g
Synonym NSC 3778 O-Methylsalicylic acid Salicylic acid methyl ether
Solubility Information DMSO : 100 mg/mL (657.25 mM; Need ultrasonic)
SMILES O=C(O)C1=CC=CC=C1OC
Molecular Weight (g/mol) 152.15
Formula Weight 152.15
Percent Purity 99.06%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.