missing translation for 'onlineSavingsMsg'
Learn More

2-Methoxybenzoic Acid, TRC

Product Code. 30133845
Change view
Click to view available options
Quantity:
1 g
10 g
50 g
Unit Size:
10g
1g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30133845 10 g 10g
30132554 50 g 50g
17962305 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30133845 Supplier Toronto Research Chemicals Supplier No. TRCM26119010G
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 579-75-9
Molecular Formula C8H8O3
Molecular Weight (g/mol) 152.15
Synonym o-Anisic acid (8CI), 2-Methoxybenzoic acid, NSC 3778, O-Methylsalicylic acid, Salicylic acid methyl ether, o-Methoxybenzoic acid, 2-Methoxybenzoic Acid
IUPAC Name 2-methoxybenzoic acid
SMILES COc1ccccc1C(=O)O

Specifications

Quantity 10 g
InChI Formula InChI=1S/C8H8O3/c1-11-7-5-3-2-4-6(7)8(9)10/h2-5H,1H3,(H,9,10)
Formula Weight 152.0473
Chemical Name or Material 2-Methoxybenzoic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.