accessibility menu, dialog, popup

UserName

H3COOHO

CAS RN 579-75-9

IUPAC Name: 2-methoxybenzoic acid
Synonyms: o-anisic acid salicylic acid methyl ether
Molecular Weight (g/mol): 152.15
Molecular Formula: C8H8O3
InChi Key: ILUJQPXNXACGAN-UHFFFAOYSA-N
SMILES: COC1=CC=CC=C1C(O)=O
Molecular Weight (g/mol) 
  • (2)
  • (7)
Quantity 
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (3)
  • (1)
Physical Form 
  • (3)
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (1)
  • (2)
  • (7)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (3)
  • (1)
  • (3)
  • (1)
  • (1)