missing translation for 'onlineSavingsMsg'
Learn More

2-Methoxybenzyl alcohol, 99%

Product Code. 10676322 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 mL
50 mL
250 mL
Packaging:
Glass bottle
Unit Size:
10mL
250mL
50mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10676322 50 mL Glass bottle 50mL
10215130 10 mL Glass bottle 10mL
10219223 250 mL Glass bottle 250mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10676322 Supplier Thermo Scientific Acros Supplier No. 125760500
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 612-16-8 | C8H10O2 | 138.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Identificatori chimici

CAS 612-16-8
Molecular Formula C8H10O2
Molecular Weight (g/mol) 138.17
MDL Number MFCD00004611
InChI Key WYLYBQSHRJMURN-UHFFFAOYSA-N
Synonym 2-methoxybenzyl alcohol, 2-methoxyphenyl methanol, o-methoxybenzyl alcohol, benzenemethanol, 2-methoxy, o-anisyl alcohol, 2-methoxybenzylalcohol, 2-methoxybenzenemethanol, unii-457nq8ngcg, 457nq8ngcg, 2-methoxyphenyl methan-1-ol
PubChem CID 69154
IUPAC Name (2-methoxyphenyl)methanol
SMILES COC1=CC=CC=C1CO

Specifica

CAS Min % 98.5
CAS Max % 100.0
Color Brown-Orange to Colorless
Density 1.0300g/mL
Boiling Point 248°C to 250°C
Flash Point >112°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3OC6H4CH2OH
Refractive Index 1.5460 to 1.548
Quantity 50 mL
Beilstein 06, 893
Specific Gravity 1.03
Formula Weight 138.17
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 2-Methoxybenzyl alcohol
Vedi altri risultati Mostra meno risultati
compliance-icons
Identificativo prodotto
  • 2-Methoxybenzyl alcohol
Avvertenza
  • Warning
Categoria di pericolo
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Indicazioni di pericolo
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Consigli di prudenza
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.