missing translation for 'onlineSavingsMsg'
Learn More

2-Methoxybenzylamine, 98%

Product Code. 10092110 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 mL
50 mL
250 mL
Packaging:
Glass bottle
Unit Size:
10mL
250mL
50mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10092110 10 mL Glass bottle 10mL
10397630 50 mL Glass bottle 50mL
10086343 250 mL Glass bottle 250mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10092110 Supplier Thermo Scientific Acros Supplier No. 170540100
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 6850-57-3 | C8H11NO | 137.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6850-57-3
Molecular Formula C8H11NO
Molecular Weight (g/mol) 137.18
MDL Number MFCD00008110
InChI Key PXJACNDVRNAFHD-UHFFFAOYSA-N
Synonym 2-methoxybenzylamine, 2-methoxyphenyl methanamine, o-methoxybenzylamine, benzenemethanamine, 2-methoxy, 2-methoxy-benzylamine, 2-methoxyphenyl methylamine, 1-2-methoxyphenyl methanamine, 2-methyoxybenzylamine, 2-methoxy benzylamine, 2-methoxybenzyl amine
PubChem CID 81292
IUPAC Name (2-methoxyphenyl)methanamine
SMILES COC1=CC=CC=C1CN

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Colorless to Yellow
Density 1.0500g/mL
Boiling Point 231°C to 233°C
Flash Point 100°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula CH3OC6H4CH2NH2
Refractive Index 1.5475 to 1.5495
Quantity 10 mL
Beilstein 13, 580
Specific Gravity 1.05
Solubility Information Solubility in water: soluble
Formula Weight 137.18
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 2-Methoxybenzylamine
Show More Show Less
compliance-icons
Product Identifier
  • 2-Methoxybenzylamine
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.