missing translation for 'onlineSavingsMsg'
Learn More

2-Methoxyhydroquinone, 97%

Product Code. 11415253
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11415253 10 g 10g
11425253 50 g 50g
11435253 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11415253 Supplier Thermo Scientific Alfa Aesar Supplier No. A12847.09
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 824-46-4 | C7H8O3 | 140.138 g/mol

It finds its application in the formation of coniferyl alcohol from ferulic acid by the white rot fungus Trametes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It finds its application in the formation of coniferyl alcohol from ferulic acid by the white rot fungus Trametes.

Solubility
Slightly soluble in water. Solubility in methanol is almost transparent.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 824-46-4
Molecular Formula C7H8O3
Molecular Weight (g/mol) 140.138
MDL Number MFCD00013971
InChI Key LAQYHRQFABOIFD-UHFFFAOYSA-N
Synonym 2-methoxyhydroquinone, methoxyhydroquinone, 1,4-benzenediol, 2-methoxy, 1,4-dihydroxy-2-methoxybenzene, unii-2hi6hnr5u1, 2hi6hnr5u1, o-methoxyhydroquinone, 2,5-dihydroxyanisole, 2-mohydrop, acmc-209pp0
PubChem CID 69988
IUPAC Name 2-methoxybenzene-1,4-diol
SMILES COC1=C(C=CC(=C1)O)O

Specifications

Melting Point 87°C to 91°C
Quantity 10 g
Beilstein 2045285
Solubility Information Slightly soluble in water. Solubility in methanol is almost transparent.
Formula Weight 140.14
Percent Purity 97%
Chemical Name or Material 2-Methoxyhydroquinone
compliance-icons
Product Identifier
  • 2-Methoxyhydroquinone
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: 2-Methoxyhydroquinone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.