missing translation for 'onlineSavingsMsg'
Learn More

2-Methoxyphenyl benzoate, 99%, Thermo Scientific™

Product Code. 11348225
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11348225 100 g 100g
11338225 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11348225 Supplier Thermo Scientific Alfa Aesar Supplier No. L08544.22
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 531-37-3
Molecular Formula C14H12O3
Molecular Weight (g/mol) 228.247
MDL Number MFCD00017153
InChI Key IZYQCDNLUPLXOO-UHFFFAOYSA-N
Synonym guaiacol benzoate, benzosol, benzoylguaiacol, o-anisyl benzoate, benzoguaiacol, benzcaine, unii-odw913za6p, guaiacyl benzoate, o-methoxyphenyl benzoate, odw913za6p
PubChem CID 68272
IUPAC Name (2-methoxyphenyl) benzoate
SMILES COC1=CC=CC=C1OC(=O)C2=CC=CC=C2

Specifications

Melting Point 57°C to 61°C
Quantity 100 g
Beilstein 2053178
Formula Weight 228.25
Percent Purity 99%
Chemical Name or Material 2-Methoxyphenyl benzoate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.