missing translation for 'onlineSavingsMsg'
Learn More

2-Methoxyphenyl isocyanate, 99%, ACROS Organics™

Product Code. 10429920 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
5g; Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10429920 5g; Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10429920 Supplier Thermo Scientific Acros Supplier No. 221150050
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 700-87-8
CAS Min % 98.5
CAS Max % 100.0
Color Colorless to Yellow or Pink
Density 1.12g/mL
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Molecular Formula C8H7NO2
Linear Formula CH3OC6H4NCO
Refractive Index 1.5404 to 1.5424 (20°C, 589nm)
MDL Number MFCD00002004
Quantity 5 g
Beilstein 13, 378
Synonym 2-methoxyphenyl isocyanate, 2-methoxyphenylisocyanate, benzene, 1-isocyanato-2-methoxy, o-methoxyphenyl isocyanate, 2-isocyanatoanisole, 1-isocyanato-2-methoxy-benzene, o-anisylisocyanate, 2-methoxybenzenisocyanate, o-anisyl isocyanate, methoxyphenyl isocyanate
InChI Key SUVCZZADQDCIEQ-UHFFFAOYSA-N
SMILES COC1=CC=CC=C1N=C=O
IUPAC Name 1-isocyanato-2-methoxybenzene
Molecular Weight (g/mol) 149.15
PubChem CID 69695
Formula Weight 149.15
Percent Purity ≥98.5%
Physical Form Liquid
Chemical Name or Material 2-Methoxyphenyl isocyanate, 99%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.