missing translation for 'onlineSavingsMsg'
Learn More

2-Methoxyphenyl isothiocyanate, 98%

Product Code. 11302627
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11302627 5 g 5g
11312627 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11302627 Supplier Thermo Scientific Alfa Aesar Supplier No. L11799.06
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 3288-04-8 | C8H7NOS | 165.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Reacts with water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents, amines ansd alcohol. It is sensitive to moisture.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3288-04-8
MDL Number MFCD00011675
ChEBI CHEBI:16318

Specifications

Density 1.196
Boiling Point 267°C
Flash Point >110°C (230°F)
Odor Pungent
Molecular Formula C6H5NO4
Refractive Index 1.6443
Quantity 5 g
UN Number UN2922
Beilstein 638293
Sensitivity Moisture sensitive
Synonym 4-nitrocatechol, 4-nitropyrocatechol, 1,2-dihydroxy-4-nitrobenzene, 4-nitro-1,2-benzenediol, 3,4-dihydroxy-1-nitrobenzene, unii-sw60ng75en, sw60ng75en, 1,2-benzenediol, 4-nitro, nitrocatechol mix of isomers, 4nc
Solubility Information Reacts with water.
InChI Key XJNPNXSISMKQEX-UHFFFAOYSA-N
SMILES C1=CC(=C(C=C1[N+](=O)[O-])O)O
IUPAC Name 4-nitrobenzene-1,2-diol
Molecular Weight (g/mol) 155.11
PubChem CID 3505109
Formula Weight 165.22
Percent Purity 98%
Chemical Name or Material 2-Methoxyphenyl isothiocyanate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.