missing translation for 'onlineSavingsMsg'
Learn More

2-Methoxyphenylacetic acid, 99%

Product Code. 11472228
Change view
Click to view available options
Quantity:
10 g
25 g
50 g
Unit Size:
10g
25g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11472228 10 g 10g
11359589 25 g 25g
11482228 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11472228 Supplier Thermo Scientific Alfa Aesar Supplier No. A15211.09
 Restricted Item
Estimated Shipment: 27-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 93-25-4 | C9H10O3 | 166.18 g/mol

2-Methoxyphenylacetic acid is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Methoxyphenylacetic acid is used as pharmaceutical intermediate.

Solubility
Soluble in water. Also soluble in methanol, ether, ethanol and acetone.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 93-25-4
Molecular Formula C9H10O3
Molecular Weight (g/mol) 166.18
MDL Number MFCD00004321
InChI Key IVEWTCACRDEAOB-UHFFFAOYSA-N
Synonym 2-methoxyphenylacetic acid, benzeneacetic acid, 2-methoxy, 2-2-methoxyphenyl acetic acid, 2-methoxyphenylaceticacid, o-methoxyphenyl acetic acid, acetic acid, o-methoxyphenyl, 2-methoxyphenyl acetic acid, o-methoxyphenylacetic acid, acmc-209rku, 2-carboxymethyl anisole
PubChem CID 7134
IUPAC Name 2-(2-methoxyphenyl)acetic acid
SMILES COC1=CC=CC=C1CC(O)=O

Specifications

Melting Point 122°C to 125°C
Quantity 10 g
Beilstein 2047573
Solubility Information Soluble in water. Also soluble in methanol,ether,ethanol and acetone.
Formula Weight 166.18
Percent Purity 99%
Chemical Name or Material 2-Methoxyphenylacetic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.