missing translation for 'onlineSavingsMsg'
Learn More

2-Methoxyphenylacetic acid, 99%

Product Code. 10052410 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 10052410

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 93-25-4 | C9H10O3 | 166.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 93-25-4
Molecular Formula C9H10O3
Molecular Weight (g/mol) 166.18
MDL Number MFCD00004321
InChI Key IVEWTCACRDEAOB-UHFFFAOYSA-N
Synonym 2-methoxyphenylacetic acid, benzeneacetic acid, 2-methoxy, 2-2-methoxyphenyl acetic acid, 2-methoxyphenylaceticacid, o-methoxyphenyl acetic acid, acetic acid, o-methoxyphenyl, 2-methoxyphenyl acetic acid, o-methoxyphenylacetic acid, acmc-209rku, 2-carboxymethyl anisole
PubChem CID 7134
IUPAC Name 2-(2-methoxyphenyl)acetic acid
SMILES COC1=CC=CC=C1CC(O)=O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 122.0°C to 125.0°C
Color Cream-Pink to White
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula CH3OC6H4CH2CO2H
Quantity 25 g
Beilstein 10, 188
Solubility Information Solubility in water: soluble. Other solubilities: soluble in ethenol,ethyl ether,acetone
Formula Weight 166.18
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material 2-Methoxyphenylacetic acid
Show More Show Less
compliance-icons
Product Identifier
  • 2-Methoxyphenylacetic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : 2-Methoxyphenylacetic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.