missing translation for 'onlineSavingsMsg'
Learn More

2-Methoxyphenylacetic acid, 99%

Product Code. 10391464 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10391464 100 g Glass bottle 100g
10052410 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10391464 Supplier Thermo Scientific Acros Supplier No. 177601000
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 93-25-4 | C9H10O3 | 166.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 93-25-4
Molecular Formula C9H10O3
Molecular Weight (g/mol) 166.18
MDL Number MFCD00004321
InChI Key IVEWTCACRDEAOB-UHFFFAOYSA-N
Synonym 2-methoxyphenylacetic acid, benzeneacetic acid, 2-methoxy, 2-2-methoxyphenyl acetic acid, 2-methoxyphenylaceticacid, o-methoxyphenyl acetic acid, acetic acid, o-methoxyphenyl, 2-methoxyphenyl acetic acid, o-methoxyphenylacetic acid, acmc-209rku, 2-carboxymethyl anisole
PubChem CID 7134
IUPAC Name 2-(2-methoxyphenyl)acetic acid
SMILES COC1=CC=CC=C1CC(O)=O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 122.0°C to 125.0°C
Color Cream-Pink to White
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula CH3OC6H4CH2CO2H
Quantity 100 g
Beilstein 10, 188
Solubility Information Solubility in water: soluble. Other solubilities: soluble in ethenol,ethyl ether,acetone
Formula Weight 166.18
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material 2-Methoxyphenylacetic acid
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.