missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-1,3-thiazole-4-carbothioamide, 97%, Thermo Scientific™

Product Code. 10213432
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
5g
Product Code. Quantity Packaging unitSize
10213432 1 g Amber Glass Bottle 1g
10275424 5 g Amber Glass Bottle 5g
2 options
This item is not returnable. View return policy

Product Code. 10213432

Brand: Thermo Scientific Maybridge SPB02802DA

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 174223-29-1
Molecular Formula C5H6N2S2
Molecular Weight (g/mol) 158.237
MDL Number MFCD00052167
InChI Key DIOPPULTIGEDCB-UHFFFAOYSA-N
Synonym 2-methylthiazole-4-carbothioamide, amino 2-methyl 1,3-thiazol-4-yl methane-1-thione, maybridge4_004334, 2-methylthiazole-4-thiocarboxamide, 4-thiazolecarbothioamide,2-methyl, 4-thiazolecarbothioamide, 2-methyl, 2-methylthiazole-4-carbothioic acid amide, 4-thiazolecarbothioamide, 2-methyl-9ci
PubChem CID 2744365
IUPAC Name 2-methyl-1,3-thiazole-4-carbothioamide
SMILES CC1=NC(=CS1)C(=S)N

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 158.25
Percent Purity 97%
Chemical Name or Material 2-Methyl-1, 3-thiazole-4-carbothioamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.