missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-1-pentene, 97%

Product Code. 15420128
Click to view available options
Quantity:
5 mL
25 mL
100 mL
Unit Size:
100mL
25mL
5mL
This item is not returnable. View return policy

Product Code. 15420128

Brand: Thermo Scientific Alfa Aesar H53373.AE

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 763-29-1 | C6H12 | 84.162 g/mol

2-Methyl-1-pentene is prepared from 2-Methyl-1-pentene. 2-methyl-1-pentene is used in copolymerization of ethylene catalyzed by nonbridged half-titanocenes. The isomerization of 2-methyl-1-pentene leads to anionic activation of c-h bonds in olefins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Methyl-1-pentene is prepared from 2-Methyl-1-pentene. 2-methyl-1-pentene is used in copolymerization of ethylene catalyzed by nonbridged half-titanocenes. The isomerization of 2-methyl-1-pentene leads to anionic activation of c-h bonds in olefins.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 763-29-1
Molecular Formula C6H12
Molecular Weight (g/mol) 84.162
MDL Number MFCD00009405
InChI Key WWUVJRULCWHUSA-UHFFFAOYSA-N
Synonym 2-methyl-1-pentene, 2-methylpentene, 1-pentene, 2-methyl, 4-methyl-4-pentene, 2-methyl-pentene-1, unii-okc25o38mc, okc25o38mc, acmc-1bixb, 4-01-00-00841 beilstein handbook reference, c2h5ch2c ch3 =ch2
PubChem CID 12986
IUPAC Name 2-methylpent-1-ene
SMILES CCCC(=C)C

Specifications

Melting Point -136°C
Density 0.682
Boiling Point 62°C
Flash Point −27°C (−17°F)
Refractive Index 1.392
Quantity 100 mL
UN Number UN2288
Beilstein 1340499
Solubility Information Insoluble in water.
Formula Weight 84.16
Percent Purity 97%
Chemical Name or Material 2-Methyl-1-pentene
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.