missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-1-tetralone, 98%

Product Code. 10627152 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
2.5 g
Packaging:
Glass bottle
Unit Size:
2.5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10627152 2.5 g Glass bottle 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10627152 Supplier Thermo Scientific Acros Supplier No. 172370025
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1590-08-5 | C11H12O | 160.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 1590-08-5
CAS Min % 97.5
CAS Max % 100.0
Color Yellow to Orange
Density 1.0600g/mL
Boiling Point 127°C to 131°C (12.0 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Molecular Formula C11H12O
Refractive Index 1.5535
MDL Number MFCD00001690
Quantity 2.5 g
Beilstein 07, I, 197
Specific Gravity 1.06
Synonym 2-methyl-1-tetralone, 2-methyl-3,4-dihydronaphthalen-1 2h-one, 1 2h-naphthalenone, 3,4-dihydro-2-methyl, 2-methyltetralin-1-one, 2-methyl-1,2,3,4-tetrahydronaphthalen-1-one, 2-methyl-2,3,4-trihydronaphthalen-1-one, 2-methyltetralone, 2-methyl-1-tetralon, acmc-20anr7, 2-methyl-alpha-tetralone
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in most organic solvents
InChI Key GANIBVZSZGNMNB-UHFFFAOYSA-N
SMILES CC1CCC2=CC=CC=C2C1=O
IUPAC Name 2-methyl-3,4-dihydro-2H-naphthalen-1-one
Molecular Weight (g/mol) 160.22
PubChem CID 102650
Formula Weight 160.22
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 2-Methyl-1-tetralone, 99%
Show More Show Less
compliance-icons
Product Identifier
  • 2-Methyl-1-tetralone
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.