missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-2-butene, 90%, balance 2-Methyl-1-butene

Product Code. 10453385 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mL
1 L
2.5 L
Packaging:
Glass bottle
Unit Size:
1 L
2.5 L
250mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10453385 1 L Glass bottle 1 L
10196673 250 mL Glass bottle 250mL
11378621 2.5 L Glass bottle 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10453385 Supplier Thermo Scientific Supplier No. 414090010
 Restricted Item
Estimated Shipment: 06-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 513-35-9 | C5H10 | 70.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 513-35-9
Molecular Formula C5H10
Molecular Weight (g/mol) 70.14
MDL Number MFCD00009276
InChI Key BKOOMYPCSUNDGP-UHFFFAOYSA-N
Synonym 2-methyl-2-butene, 2-butene, 2-methyl, trimethylethylene, amylene, n-amylene, 3-methyl-2-butene, ethylene, trimethyl, beta-isoamylene, 1,1,2-trimethylethylene, 2-methylbutene-2
PubChem CID 10553
ChEBI CHEBI:77916
IUPAC Name 2-methylbut-2-ene
SMILES CC=C(C)C

Specifications

Melting Point -134.0°C
Color Colorless to Yellow
Density 0.6600g/mL
Boiling Point 35.0°C to 38.0°C
Flash Point −45°C
Infrared Spectrum Authentic
Assay Percent Range Balance mainly 2-methyl-1-butene (GC)
Packaging Glass bottle
Linear Formula CH3CH=C(CH3)2
Refractive Index 1.3860 to 1.3880
Quantity 1 L
Beilstein 01,211
Merck Index 15,601
Specific Gravity 0.66
Formula Weight 70.13
Percent Purity 90%
Physical Form Liquid
Chemical Name or Material 2-Methyl-2-butene
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.