missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-3,5-dinitrobenzoic acid, 97+%

Product Code. 11491938
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11491938 25 g 25g
11401948 100 g 100g
11329589 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11491938 Supplier Thermo Scientific Alfa Aesar Supplier No. A14555.14
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 28169-46-2 | C8H6N2O6 | 226.14 g/mol

2-Methyl-3,5-dinitrobenzoic acid was used to synthesis heterocyclic derivatives and in the synthesis of isoquinolone.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Methyl-3,5-dinitrobenzoic acid was used to synthesis heterocyclic derivatives and in the synthesis of isoquinolone.

Solubility
Soluble in water.

Notes
Store in cool, dry conditions in a well sealed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 28169-46-2
Molecular Formula C8H6N2O6
Molecular Weight (g/mol) 226.14
MDL Number MFCD00007161
InChI Key CDVNZMKTJIBBBV-UHFFFAOYSA-N
Synonym 2-methyl-3,5-dinitrobenzoic acid, 3,5-dinitro-2-methylbenzoic acid, 3,5-dinitro-o-toluic acid, benzoic acid, 2-methyl-3,5-dinitro, 3,5-dinitrotoluic acid, o-toluic acid, 3,5-dinitro, 2-methyl-3,5-dinitro-benzoic acid, 3,5-dinitro-2-methyl-benzoic acid, acmc-1cfeu, o-toluic acid,5-dinitro
PubChem CID 98824
IUPAC Name 2-methyl-3,5-dinitrobenzoic acid
SMILES CC1=C(C=C(C=C1[N+]([O-])=O)[N+]([O-])=O)C(O)=O

Specifications

Melting Point 203°C to 207°C
Quantity 25 g
Beilstein 2622906
Solubility Information Soluble in water.
Formula Weight 226.15
Percent Purity 98%
Chemical Name or Material 2-Methyl-3,5-dinitrobenzoic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.