missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-3-butyn-2-ol, 98%

Product Code. 11924621 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mL
1 L
2.5 L
Packaging:
Glass bottle
Unit Size:
1 L
2.5 L
250mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
11924621 2.5 L Glass bottle 2.5 L
10502561 250 mL Glass bottle 250mL
10449260 1 L Glass bottle 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
Product Code. 11924621 Supplier Thermo Scientific Acros Supplier No. 156820025
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

CAS: 115-19-5 | C5H8O | 84.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemische Identifikatoren

CAS 115-19-5
Molecular Formula C5H8O
Molecular Weight (g/mol) 84.12
InChI Key CEBKHWWANWSNTI-UHFFFAOYSA-N
Synonym 2-methyl-3-butyn-2-ol, 3-butyn-2-ol, 2-methyl, 3-methyl-1-butyn-3-ol, 3-methylbutynol, 2-methyl-2-butynol, ethynyldimethylcarbinol, carbavane, dimethylethynylcarbinol, dimethylethynylmethanol, 1,1-dimethyl-2-propynol
PubChem CID 8258
IUPAC Name 2-methylbut-3-yn-2-ol
SMILES CC(C)(C#C)O

Spezifikation

Melting Point 3.0°C
Color Brown to White
Density 0.8608g/mL
Boiling Point 104.0°C
Flash Point 21°C
Infrared Spectrum Authentic
Assay Percent Range 1% max. tetrahydrofuran (GC)
Packaging Glass bottle
Linear Formula HC≡CC(CH3)2OH
Refractive Index 1.4200 to 1.4220
Quantity 2.5 L
Beilstein 01,I,235
Fieser 01,674
Merck Index 15,6108
Specific Gravity 0.8608
Solubility Information Solubility in water: miscible. Other solubilities: miscible with acetone,benzene,cellosolve,,carbon tetrachloride,cyclohexanone,diethylene,glycol,ethyl acetate,kerosine,methyl ethyl,ketone,mineral spirits,monoethanolamine
Viscosity 3.02 mPa.s (20°C)
Formula Weight 84.12
Percent Purity 98%
Physical Form Crystals or Powder
Chemical Name or Material 2-Methyl-3-butyn-2-ol
Mehr anzeigen Weniger anzeigen
compliance-icons compliance-icons compliance-icons compliance-icons
Produktidentifikator
  • 2-Methyl-3-butyn-2-ol
Signalwort
  • Danger
Gefahrenkategorie
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Specific target organ toxicity Category 3
  • Reproductive toxicity Category 2
Gefahrenhinweis
  • H318-Causes serious eye damage.
  • H336-May cause drowsiness or dizziness.
  • H302-Harmful if swallowed.
  • H225-Highly flammable liquid and vapour.
  • H361f-Suspected of damaging fertility.
Sicherheitshinweis
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P264-Wash face, hands and any thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.