missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-3-nitrobenzotrifluoride, 97%

Product Code. 11469157
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11469157 25 g 25g
11444807 1 g 1g
11459157 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11469157 Supplier Thermo Scientific Alfa Aesar Supplier No. B21267.14
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 6656-49-1 | C8H6F3NO2 | 205.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6656-49-1
Molecular Formula C8H6F3NO2
Molecular Weight (g/mol) 205.14
MDL Number MFCD00042322
InChI Key KQUQBPVYIURTNZ-UHFFFAOYSA-N
Synonym 2-methyl-3-nitrobenzotrifluoride, 2-nitro-6-trifluoromethyltoluene, 2-methyl-1-nitro-3-trifluoromethyl benzene, 2-nitro-6-trifluoromethyl toluene, 2-methyl-3-trifluoromethyl nitrobenzene, benzene, 2-methyl-1-nitro-3-trifluoromethyl, alpha,alpha,alpha-trifluoro-3-nitro-o-xylene, 3-trifluoromethyl-2-methyl-1-nitrobenzene, pubchem4816, 2-methyl-3-nitro-benzotrifluoride
PubChem CID 81171
IUPAC Name 2-methyl-1-nitro-3-(trifluoromethyl)benzene
SMILES CC1=C(C=CC=C1C(F)(F)F)[N+]([O-])=O

Specifications

Boiling Point 86°C (10 mmHg)
Flash Point >100°C (212°F)
Refractive Index 1.478
Quantity 25 g
UN Number UN2810
Beilstein 2457216
Sensitivity Light sensitive
Formula Weight 205.14
Percent Purity 97%
Chemical Name or Material 2-Methyl-3-nitrobenzotrifluoride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.