missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Methyl-3-nitrobenzyl chloride, 97%, Thermo Scientific™
Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Specifications
Specifications
| CAS Min % | 96.0 |
| CAS Max % | 100.0 |
| Melting Point | 42°C to 47°C |
| Flash Point | >109°C |
| Assay Percent Range | 96% min. (GC) |
| Linear Formula | CH3C6H3(NO2)CH2Cl |
| Quantity | 5 g |
| Formula Weight | 185.61 |
| Percent Purity | 97% |
| Chemical Name or Material | 2-Methyl-3-nitrobenzyl chloride, 97% |
Safety and Handling
Product Identifier
- 2-Methyl-3-nitrobenzyl chloride
- Danger
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction