missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-3-nitrobenzyl chloride, 97%, Thermo Scientific™

Product Code. p-164963 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 10376641

Brand: Thermo Scientific Acros 187400010

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 60468-54-4
Molecular Formula C8H8ClNO2
Molecular Weight (g/mol) 185.61
MDL Number MFCD00007165
InChI Key XZNDXQGZPOZITR-UHFFFAOYSA-N
Synonym 2-methyl-3-nitrobenzyl chloride, 1-chloromethyl-2-methyl-3-nitrobenzene, alpha-chloro-2-methyl-3-nitrotoluene, 1-chloromethyl-2-methyl-3-nitro-benzene, 2-methyl-3-nitrobenzylchloride, benzene, 1-chloromethyl-2-methyl-3-nitro, acmc-1b2j5, ksc497o8t, 2-chloromethyl-6-nitrotoluene
PubChem CID 108947
IUPAC Name 1-(chloromethyl)-2-methyl-3-nitrobenzene
SMILES CC1=C(CCl)C=CC=C1[N+]([O-])=O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 42°C to 47°C
Flash Point >109°C
Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Linear Formula CH3C6H3(NO2)CH2Cl
Quantity 1 g
Formula Weight 185.61
Percent Purity 97%
Chemical Name or Material 2-Methyl-3-nitrobenzyl chloride, 97%
Show More Show Less
compliance-icons
Product Identifier
  • 2-Methyl-3-nitrobenzyl chloride
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.