missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-3-nitrophenylacetic acid, 96%

Product Code. 15482179
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15482179 25 g 25g
15472179 5 g 5g
15492179 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15482179 Supplier Thermo Scientific Alfa Aesar Supplier No. H64810.14
 Restricted Item
Estimated Shipment: 21-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 23876-15-5 | C9H9NO4 | 195.174 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 23876-15-5
Molecular Formula C9H9NO4
Molecular Weight (g/mol) 195.174
MDL Number MFCD07782132
InChI Key LWIOFILTAJJDLA-UHFFFAOYSA-N
Synonym 2-methyl-3-nitrophenylacetic acid, 2-methyl-3-nitrophenyl acetic acid, 2-2-methyl-3-nitrophenyl acetic acid, 2-methyl-3-nitro-benzeneacetic acid, 3-nitro-o-tolyl acetic acid, benzeneacetic acid, 2-methyl-3-nitro, 2-methyl-3-nitrophenylaceticacid, 2-methyl-3-nitrobenzeneacetic acid, 2-methyl-3-nitro phenyl acetic acid, pubchem18230
PubChem CID 13530897
SMILES CC1=C(C=CC=C1[N+](=O)[O-])CC(=O)O

Specifications

Melting Point 132°C to 136°C
Quantity 25 g
Beilstein 2581973
Formula Weight 195.17
Percent Purity 96%
Physical Form Solid
Chemical Name or Material 2-Methyl-3-nitrophenylacetic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.