missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-3-phenyl-1-propene, 99%

Product Code. 11454277
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11454277 5 g 5g
11464277 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11454277 Supplier Thermo Scientific Alfa Aesar Supplier No. B20370.06
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 3290-53-7 | C10H12 | 132.206 g/mol

It is used as an active pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as an active pharmaceutical intermediate.

Solubility
Slighlty soluble in aqeous solvents.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3290-53-7
Molecular Formula C10H12
Molecular Weight (g/mol) 132.206
MDL Number MFCD00039815
InChI Key MXTNFIYGTWARIN-UHFFFAOYSA-N
Synonym 2-methyl-3-phenyl-1-propene, methallylbenzene, benzene, 2-methyl-2-propenyl, benzene, 2-methylallyl, 2-methylprop-2-en-1-yl benzene, 2-methyl-2-propenyl benzene, methallyl benzene, 2-methylallylbenzene, 2-benzyl-1-propene, 2-methylallyl benzene
PubChem CID 18687
IUPAC Name 2-methylprop-2-enylbenzene
SMILES CC(=C)CC1=CC=CC=C1

Specifications

Quantity 5 g
Solubility Information Slighlty soluble in aqeous solvents.
Formula Weight 132.21
Percent Purity 99%
Chemical Name or Material 2-Methyl-3-phenyl-1-propene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.