missing translation for 'onlineSavingsMsg'
Learn More

2'-Methyl-4'-nitroacetanilide, 97%, Thermo Scientific™

Product Code. 11489547
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11489547

Brand: Thermo Scientific Alfa Aesar B22054.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
2′-Methyl-4′-nitroacetanilide acts as the reactant in the synthesis of TNT isomers.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, acids, bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2719-15-5
Molecular Formula C9H10N2O3
Molecular Weight (g/mol) 194.19
MDL Number MFCD00033896
InChI Key JZEOVPGWIWSSAK-UHFFFAOYSA-N
Synonym n-2-methyl-4-nitrophenyl acetamide, 4'-nitro-o-acetotoluidide, n1-2-methyl-4-nitrophenyl acetamide, acetamide, n-2-methyl-4-nitrophenyl, 2'-methyl-4'-nitroacetanilide, acetyl-p-nitro-o-toluidine, 2-methyl-4-nitroacetanilide, o-acetotoluidide, 4'-nitro, acetamide,n-2-methyl-4-nitrophenyl, maybridge1_001434
PubChem CID 75936
IUPAC Name N-(2-methyl-4-nitrophenyl)acetamide
SMILES CC(=O)NC1=CC=C(C=C1C)[N+]([O-])=O

Specifications

Melting Point 198°C to 200°C
Quantity 25 g
Formula Weight 194.19
Percent Purity 97%
Chemical Name or Material 2'-Methyl-4'-nitroacetanilide
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.