missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-4-nitrobenzoic acid, 97%

Product Code. 15464067
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15464067 1 g 1g
15474067 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15464067 Supplier Thermo Scientific Alfa Aesar Supplier No. H31332.03
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1975-51-5 | C8H7NO4 | 181.147 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1975-51-5
Molecular Formula C8H7NO4
Molecular Weight (g/mol) 181.147
MDL Number MFCD00210697
InChI Key XXXOBNJIIZQSPT-UHFFFAOYSA-N
Synonym 2-methyl-4-nitrobenzoic acid, 4-nitro-o-toluic acid, 2-methyl-4-nitro-benzoic acid, 2-methyl-4-nitrobenzoicacid, 4-nitro-2-methylbenzoic acid, benzoic acid, 2-methyl-4-nitro, pubchem8363, acmc-1bsab, 2-carboxy-5-nitrotoluene, 2-methyl4-nitrobenzoic acid
PubChem CID 74790
SMILES CC1=C(C=CC(=C1)[N+](=O)[O-])C(=O)O

Specifications

Melting Point 152°C to 155°C
Odor Odorless
Quantity 1 g
Formula Weight 181.15
Percent Purity 97%
Chemical Name or Material 2-Methyl-4-nitrobenzoic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.