missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-4-nitrobenzoic acid, 98%

Product Code. 15535276 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 15535276

Brand: Thermo Scientific Acros 457810250

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1975-51-5 | C8H7NO4 | 181.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1975-51-5
Molecular Formula C8H7NO4
Molecular Weight (g/mol) 181.15
InChI Key XXXOBNJIIZQSPT-UHFFFAOYSA-N
Synonym 2-methyl-4-nitrobenzoic acid, 4-nitro-o-toluic acid, 2-methyl-4-nitro-benzoic acid, 2-methyl-4-nitrobenzoicacid, 4-nitro-2-methylbenzoic acid, benzoic acid, 2-methyl-4-nitro, pubchem8363, acmc-1bsab, 2-carboxy-5-nitrotoluene, 2-methyl4-nitrobenzoic acid
PubChem CID 74790
SMILES CC1=C(C=CC(=C1)[N+](=O)[O-])C(=O)O

Specifications

Melting Point 151.0°C to 152.0°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Quantity 25 g
Formula Weight 181.15
Percent Purity 98%
Chemical Name or Material 2-Methyl-4-nitrobenzoic acid
compliance-icons
Product Identifier
  • 2-Methyl-4-nitrobenzoic acid, 98%
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.