missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-Methyl-5-nitrobenzeneboronic acid, 96%

Product Code. 15499917
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15499917 1 g 1g
15409927 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15499917 Supplier Alfa Aesar™ Supplier No. H53160.03
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 100960-11-0
Molecular Formula C7H8BNO4
Molecular Weight (g/mol) 180.954
MDL Number MFCD00757435
InChI Key HRJFQIRIDFXMNX-UHFFFAOYSA-N
Synonym 2-methyl-5-nitrophenyl boronic acid, 2-methyl-5-nitrobenzeneboronic acid, 2-methyl-5-nitrophenyl boranediol, boronic acid,b-2-methyl-5-nitrophenyl, 5-nitro-2-methyl-phenylboronic acid, 2-dimethyl-3-nitrophenylboronic acid, 2-borono-4-nitrotoluene, acmc-2097th, 3-borono-4-methylnitrobenzene, 2-methyl-5-nitrophenyl-boronic acid
PubChem CID 2773513
IUPAC Name (2-methyl-5-nitrophenyl)boronic acid
SMILES B(C1=C(C=CC(=C1)[N+](=O)[O-])C)(O)O

Specifications

Melting Point 62°C to 66°C
Quantity 1 g
Formula Weight 180.96
Percent Purity 96%
Chemical Name or Material 2-Methyl-5-nitrobenzeneboronic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.