missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-5-tert-butylthiophenol, 93%, remainder isomer

Product Code. 10712301 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
50 g
Packaging:
Glass bottle
Unit Size:
50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10712301 50 g Glass bottle 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10712301 Supplier Thermo Scientific Acros Supplier No. 279960500
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 7340-90-1 | C11H16S | 180.31 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 7340-90-1
CAS Min % 91.0
CAS Max % 100.0
Melting Point -10°C
Color Colorless
Density 0.9800g/mL
Boiling Point 260°C
Flash Point 123°C
Infrared Spectrum Authentic
Assay Percent Range 93%
Packaging Glass bottle
Molecular Formula C11H16S
Refractive Index 1.5460 to 1.548
MDL Number MFCD00142942
Quantity 50 g
Specific Gravity 0.98
Synonym 5-tert-butyl-2-methylthiophenol, 4-tert-butyl-2-mercaptotoluene, 5-tert-butyl-o-thiocresol, benzenethiol, 5-1,1-dimethylethyl-2-methyl, acmc-1bejs, zsbnxoiajfvxmp-uhfffaoysa, 2-methyl-5-tert-butyl thiophenol, 2-methyl-5-tert-butylbenzenethiol, 2-methyl-5-tert.-butyl-thiophenol, 5-tert-butyl-2-methylbenzenethiol
Solubility Information Solubility in water: 0.0035g/L (20°C). Other solubilities: miscible with n-heptane,methanol,ethanol and,isopropanol
InChI Key ZSBNXOIAJFVXMP-UHFFFAOYSA-N
SMILES CC1=C(C=C(C=C1)C(C)(C)C)S
IUPAC Name 5-tert-butyl-2-methylbenzenethiol
Molecular Weight (g/mol) 180.31
PubChem CID 596868
Formula Weight 180.31
Percent Purity 93%
Physical Form Liquid
Chemical Name or Material 2-Methyl-5-tert-butylthiophenol, 93%
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 2-Methyl-5-tert-butylthiophenol, remainder isomer
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • Aspiration toxicity Category 1
  • Serious eye damage/eye irritation Category 2
  • Reproductive toxicity Category 2
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity after repeated exposure Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H361d-Suspected of damaging the unborn child.
  • H373-May cause damage to organs.
  • H336-May cause drowsiness or dizziness.
  • H304-May be fatal if swallowed and enters airways.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.