missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-5-(trifluoromethoxy)benzylamine, 96%, Thermo Scientific Chemicals

Product Code. 15496647
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15496647 1 g 1g
15406657 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15496647 Supplier Thermo Scientific Alfa Aesar Supplier No. H33791.03
 Restricted Item
Estimated Shipment: 17-08-2026
Add to basket
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1373920-87-6
Molecular Formula C9H10F3NO
Molecular Weight (g/mol) 205.18
MDL Number MFCD22201056
InChI Key HUHITEUBBNSNER-UHFFFAOYSA-N
Synonym 2-methyl-5-trifluoromethoxy benzylamine, 2-methyl-5-trifluoromethoxy phenyl methanamine, 1-2-methyl-5-trifluoromethoxy phenyl methanamine
PubChem CID 86277657
IUPAC Name [2-methyl-5-(trifluoromethoxy)phenyl]methanamine
SMILES CC1=C(CN)C=C(OC(F)(F)F)C=C1

Specifications

Quantity 1 g
UN Number UN3267
Sensitivity Air sensitive
Formula Weight 205.18
Percent Purity 96%
Chemical Name or Material 2-Methyl-5-(trifluoromethoxy)benzylamine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.