missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-6-nitroaniline 99%, ACROS Organics™

Product Code. 10606724 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10606724 100 g Glass bottle 100g
10294980 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10606724 Supplier Thermo Scientific Acros Supplier No. 127231000
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 570-24-1
Molecular Formula C7H8N2O2
Molecular Weight (g/mol) 152.153
MDL Number MFCD00007744
InChI Key FCMRHMPITHLLLA-UHFFFAOYSA-N
Synonym 6-nitro-o-toluidine, 2-amino-3-nitrotoluene, benzenamine, 2-methyl-6-nitro, 2-methyl-6-nitrobenzenamine, 6-methyl-2-nitroaniline, 1-amino-2-methyl-6-nitrobenzene, unii-oup165ykbc, 3-nitro-2-aminotoluene, 2-nitro-6-methylaniline, oup165ykbc
PubChem CID 11298
IUPAC Name 2-methyl-6-nitroaniline
SMILES CC1=C(C(=CC=C1)[N+](=O)[O-])N

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 93.0°C to 96.0°C
Color Brown
Flash Point 110°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula CH3C6H3(NO2)NH2
Quantity 100 g
Beilstein 12, 843
Solubility Information Solubility in water: negligible. Other solubilities: slouble in ethanol and ether
Formula Weight 152.15
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material 2-Methyl-6-nitroaniline, 99%
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 2-Methyl-6-nitroaniline
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H373-May cause damage to organs.
  • H411-Toxic to aquatic life with long lasting effects.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.