missing translation for 'onlineSavingsMsg'
Learn More

(±)-2-Methyl-d3-succinic-2,3,3-d3 Acid, TRC

Product Code. 30478192
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30478192 50 mg 50mg
30483137 10 mg 10mg
30466120 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30478192 Supplier Toronto Research Chemicals Supplier No. TRCM32904650MG
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 347840-08-8
Molecular Formula C5 D6 H2 O4
Molecular Weight (g/mol) 138.152
Synonym Butanedioic-d3 acid, methyl-d3- (9CI), Methyl-d3-butanedioic-d3 acid, (±)-2-Methyl-d3-succinic-2,3,3-d3 Acid, Methylsuccinic Acid-D6, 2-(Methyl-d3)butanedioic-2,3,3-d3 acid, 2-(Methyl-d3)succinic-2,3,3-d3 acid
IUPAC Name 2,2,3-trideuterio-3-(trideuteriomethyl)butanedioic acid
SMILES [2H]C([2H])([2H])C([2H])(C(=O)O)C([2H])([2H])C(=O)O

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C5H8O4/c1-3(5(8)9)2-4(6)7/h3H,2H2,1H3,(H,6,7)(H,8,9)/i1D3,2D2,3D
Formula Weight 138.08
Chemical Name or Material (±)-2-Methyl-d3-succinic-2,3,3-d3 Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.