missing translation for 'onlineSavingsMsg'
Learn More

2'-Methylacetanilide, 98+%

Product Code. 11409040
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11409040 25 g 25g
11419040 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11409040 Supplier Thermo Scientific Alfa Aesar Supplier No. A13235.14
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 120-66-1 | C9H11NO | 149.193 g/mol

Acetanilide is used as an inhibitor of peroxides and stabilizer for cellulose ester varnishes. It is used as an intermediate for the synthesis of rubber accelerators, dyes and dye intermediate and camphor. It is used as a precursor in penicillin synthesis and other pharmaceuticals including painkillers and intermediates. 

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Acetanilide is used as an inhibitor of peroxides and stabilizer for cellulose ester varnishes. It is used as an intermediate for the synthesis of rubber accelerators, dyes and dye intermediate and camphor. It is used as a precursor in penicillin synthesis and other pharmaceuticals including painkillers and intermediates. 

Solubility
It is slightly soluble in acetone, chloroform, toluene, benzene.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from Strong oxidizing agents. Stable under ordinary conditions.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 120-66-1
Molecular Formula C9H11NO
Molecular Weight (g/mol) 149.193
MDL Number MFCD00014961
InChI Key BPEXTIMJLDWDTL-UHFFFAOYSA-N
Synonym o-acetotoluidide, 2'-methylacetanilide, n-o-tolyl acetamide, n-2-methylphenyl acetamide, acetyl-o-toluidine, n-acetyl-o-toluidine, o-acetotoluide, acetamide, n-2-methylphenyl, o-methylacetanilide, n-o-tolylacetamide
PubChem CID 8443
IUPAC Name N-(2-methylphenyl)acetamide
SMILES CC1=CC=CC=C1NC(=O)C

Specifications

Melting Point 109°C to 112°C
Density 1.16
Boiling Point 296°C
Quantity 25 g
Beilstein 2207054
Merck Index 14,74
Solubility Information It is slightly soluble in acetone,chloroform,toluene,benzene.
Formula Weight 149.19
Percent Purity ≥98%
Chemical Name or Material 2'-Methylacetanilide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.