missing translation for 'onlineSavingsMsg'
Learn More

2-(Methylamino)-1H-purin-6(7H)-one, MedChemExpress

Product Code. 16476077
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16476077 5 mg 5mg
16464596 100 mg 100mg
16429886 50 mg 50mg
16403358 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16476077 Supplier Medchem Express Supplier No. HY101412
 Restricted Item
Estimated Shipment: 07-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress 2-(Methylamino)-1H-purin-6(7H)-one (N2-Methylguanine) is a modified nucleoside. 2-(Methylamino)-1H-purin-6(7H)-one is an endogenous methylated nucleoside found in human fluids.

2-(Methylamino)-1H-purin-6(7H)-one (N2-Methylguanine) is a modified nucleoside. 2-(Methylamino)-1H-purin-6(7H)-one is an endogenous methylated nucleoside found in human fluids.

Specifications

Chemical Name or Material 2-(Methylamino)-1H-purin-6(7H)-one
Physical Form Solid
CAS 10030-78-1
Color Earth Yellow
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C6H7N5O
Synonym N2-methylguanine
Solubility Information H2O : 1 mg/mL (6.06 mM; ultrasonic and warming and heat to 80°C) ∣DMSO : 1 mg/mL (6.06 mM; Need ultrasonic) ∣Ethanol : < 1 mg/mL (ultrasonic) (insoluble) ∣THF : < 1 mg/mL (ultrasonic) (insoluble) ∣1M NaOH : < 1 mg/mL (ultrasonic;adjust pH to 12 with NaOH) (insoluble) ∣Acetone : < 1 mg/mL (ultrasonic) (insoluble) ∣0.1 M HCL : < 1 mg/mL (ultrasonic;adjust pH to 2 with HCl) (insoluble) ∣DMF : < 1 mg/mL (ultrasonic) (insoluble)
SMILES O=C1N=C(NC)NC2=C1NC=N2
Molecular Weight (g/mol) 165.15
Formula Weight 165.15
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.