missing translation for 'onlineSavingsMsg'
Learn More

2-Methylbenzothiazol-5-ol, 97%

Product Code. 15405727
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15405727 1 g 1g
15415727 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15405727 Supplier Thermo Scientific Alfa Aesar Supplier No. H32937.03
 Restricted Item
Estimated Shipment: 26-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 68867-14-1 | C8H7NOS | 165.21 g/mol

Used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used as pharmaceutical intermediate.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 68867-14-1
Molecular Formula C8H7NOS
Molecular Weight (g/mol) 165.21
MDL Number MFCD00192276
InChI Key LAKVUPMDDFICNR-UHFFFAOYSA-N
PubChem CID 699799
IUPAC Name 2-methyl-1,3-benzothiazol-5-ol
SMILES CC1=NC2=CC(O)=CC=C2S1

Specifications

Melting Point 185°C to 187°C
Quantity 1 g
Synonym 2-methylbenzo d thiazol-5-ol, 5-benzothiazolol, 2-methyl, 2-methyl-5-benzothiazolol, 2-methylbenzothiazol-5-ol, 5-benzothiazolol,2-methyl, 5-hydroxy-2-methylbenzothiazole, 2-methyl-benzothiazol-5-ol, 2-methyl-5-hydroxybenzothiazole
Formula Weight 165.21
Percent Purity 97%
Chemical Name or Material 2-Methylbenzothiazol-5-ol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.