missing translation for 'onlineSavingsMsg'
Learn More

2-Methylbenzyl chloride, 98+%

Product Code. 11415513
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
This item is not returnable. View return policy

Product Code. 11415513

Brand: Thermo Scientific Alfa Aesar A13448.36

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 552-45-4 | C8H9Cl | 140.61 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 552-45-4
Molecular Formula C8H9Cl
Molecular Weight (g/mol) 140.61
MDL Number MFCD00000899
InChI Key VQRBXYBBGHOGFT-UHFFFAOYSA-N
Synonym 2-methylbenzyl chloride, 1-chloromethyl-2-methylbenzene, o-xylyl chloride, alpha-chloro-o-xylene, o-methylbenzyl chloride, o-chloromethyl toluene, benzene, 1-chloromethyl-2-methyl, 2-methylbenzylchloride, 2-chloromethyl toluene, unii-eqt3v044tw
PubChem CID 11093
IUPAC Name 1-(chloromethyl)-2-methylbenzene
SMILES CC1=CC=CC=C1CCl

Specifications

Melting Point 2°C
Density 1.081
Boiling Point 197°C to 199°C
Flash Point 73°C (163°F)
Odor Pungent
Refractive Index 1.541
Quantity 500 g
UN Number UN3265
Beilstein 386016
Merck Index 14,10091
Formula Weight 140.61
Percent Purity ≥98%
Chemical Name or Material 2-Methylbenzyl chloride
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.