missing translation for 'onlineSavingsMsg'
Learn More

2-Methylbenzylamine, 98%

Product Code. 11397355
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11397355 1 g 1g
11307365 5 g 5g
11317365 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11397355 Supplier Thermo Scientific Alfa Aesar Supplier No. L05243.03
 Restricted Item
Estimated Shipment: 30-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 89-93-0 | C8H11N | 121.183 g/mol

2-Methylbenzylamine, is used in the preparation of boc-2-alkylanilines: N- (tert-butoxycarbonyl)-1-methylbenzylamine

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Methylbenzylamine, is used in the preparation of boc-2-alkylanilines: N- (tert-butoxycarbonyl)-1-methylbenzylamine

Solubility
Very soluble in water.

Notes
Air Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents, strong acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 89-93-0
Molecular Formula C8H11N
Molecular Weight (g/mol) 121.183
MDL Number MFCD00008112
InChI Key CJAAPVQEZPAQNI-UHFFFAOYSA-N
Synonym 2-methylbenzylamine, o-tolylmethanamine, o-methylbenzylamine, benzenemethanamine, 2-methyl, o-xylylamine, benzylamine, o-methyl, 2-methylphenyl methanamine, alpha-amino-o-xylene, alpha-amino-2-xylene, 1-2-methylphenyl methanamine
PubChem CID 6993
IUPAC Name (2-methylphenyl)methanamine
SMILES CC1=CC=CC=C1CN

Specifications

Boiling Point 198°C to 200°C
Flash Point 83°C (181°F)
Quantity 1 g
UN Number UN2735
Beilstein 1099406
Formula Weight 121.18
Percent Purity 98%
Chemical Name or Material 2-Methylbenzylamine
compliance-icons
Product Identifier
  • 2-Methylbenzylamine
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: 2-Methylbenzylamine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.