missing translation for 'onlineSavingsMsg'
Learn More

2-Methylene-1,3-propanediol, 97%

Product Code. 10441881 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 10441881

Brand: Thermo Scientific Acros 308590010

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 3513-81-3 | C4H8O2 | 88.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3513-81-3
Molecular Formula C4H8O2
Molecular Weight (g/mol) 88.11
MDL Number MFCD00075162
InChI Key JFFYKITVXPZLQS-UHFFFAOYSA-N
Synonym 2-methylene-1,3-propanediol, 2-methylenepropane-1,3-diol, 1,3-propanediol, 2-methylene, 1,3-dihydroxy-2-methylenepropane, 1,3-dihydroxy-2-methylidenepropane, dimethylol ethylene, 2-methylene1,3-propanediol, 2-methylenepropan-1,3-diol, ksc573o4t, 1,3-propanediol,2-methylene
PubChem CID 77048
IUPAC Name 2-methylidenepropane-1,3-diol
SMILES C=C(CO)CO

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Colorless to Yellow
Density 1.0800g/mL
Boiling Point 93°C to 95°C (2.0 mmHg)
Flash Point <110°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula HOCH2C(=CH2)CH2OH
Refractive Index 1.4730 to 1.475
Quantity 1 g
Specific Gravity 1.08
Formula Weight 88.11
Percent Purity 97%
Physical Form Viscous Liquid
Chemical Name or Material 2-Methylene-1, 3-propanediol
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.