missing translation for 'onlineSavingsMsg'
Learn More

2-Methylhydroquinone, 99%

Product Code. p-7025638
Click to view available options
Quantity:
250 g
1000 g
Unit Size:
1000g
250g
This item is not returnable. View return policy

Product Code. 11414767

Brand: Thermo Scientific Alfa Aesar B21241.30

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 95-71-6 | C7H8O2 | 124.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 95-71-6
Molecular Formula C7H8O2
Molecular Weight (g/mol) 124.14
MDL Number MFCD00002345
InChI Key CNHDIAIOKMXOLK-UHFFFAOYSA-N
Synonym methylhydroquinone, 2-methylhydroquinone, 2,5-dihydroxytoluene, p-toluhydroquinone, toluhydroquinone, toluquinol, p-toluquinol, tolylhydroquinone, p-toluhydroquinol, 2,5-toluenediol
PubChem CID 7253
IUPAC Name 2-methylbenzene-1,4-diol
SMILES CC1=CC(O)=CC=C1O

Specifications

Melting Point 127°C to 130°C
Boiling Point 284°C to 286°C
Flash Point 172°C (341°F)
Quantity 250 g
Beilstein 2041489
Formula Weight 124.14
Percent Purity 99%
Chemical Name or Material 2-Methylhydroquinone

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.