missing translation for 'onlineSavingsMsg'
Learn More

2-Methylphenoxyacetic acid, 98%

Product Code. 11464394
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11464394 250 g 250g
11444394 10 g 10g
11454394 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11464394 Supplier Thermo Scientific Alfa Aesar Supplier No. A13076.30
 Restricted Item
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1878-49-5 | C9H10O3 | 166.176 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1878-49-5
Molecular Formula C9H10O3
Molecular Weight (g/mol) 166.176
MDL Number MFCD00014354
InChI Key QJVXBRUGKLCUMY-UHFFFAOYSA-N
Synonym 2-methylphenoxy acetic acid, 2-2-methylphenoxy acetic acid, 2-methylphenoxyacetic acid, acetic acid, 2-methylphenoxy, o-tolyloxyacetic acid, o-tolyloxy acetic acid, 2-o-tolyloxy acetic acid, acetic acid, o-tolyloxy, acide o-methylphenoxyacetique, unii-pxo85n85g3
PubChem CID 74651
IUPAC Name 2-(2-methylphenoxy)acetic acid
SMILES CC1=CC=CC=C1OCC(=O)O

Specifications

Melting Point 152°C to 156°C
Quantity 250 g
Beilstein 1943465
Formula Weight 166.18
Percent Purity 98%
Chemical Name or Material 2-Methylphenoxyacetic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.