missing translation for 'onlineSavingsMsg'
Learn More

2-Methylpropanimidamide hydrochloride, ≥95%, Thermo Scientific™

Product Code. 10407822
Change view
Click to view available options
Quantity:
1 g
10 g
50 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10407822 1 g Amber Glass Bottle 1g
10541132 10 g Amber Glass Bottle 10g
10059381 50 g Amber Glass Bottle 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10407822 Supplier Thermo Scientific Maybridge Supplier No. RF02291DA
 Restricted Item
Estimated Shipment: 20-11-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 22007-68-7
Molecular Formula C4H11ClN2
Molecular Weight (g/mol) 122.596
MDL Number MFCD00173787
InChI Key VWXLCWNPSOUPPE-UHFFFAOYSA-N
Synonym 2-methylpropanimidamide hydrochloride, isobutyrimidamide hydrochloride, isobutyramidine hydrochloride, 2-methylpropionamidine hcl, isobutanimidamide hydrochloride, isopropylcarbamidine hydrochloride, 2-methylpropionamidine hydrochloride, 2-methyl propanimidamide hydrochloride, isobutanimidamide hcl, sothiadhk\up@
PubChem CID 2782053
IUPAC Name 2-methylpropanimidamide;hydrochloride
SMILES CC(C)C(=N)N.Cl

Specifications

CAS Min % 90.0
Assay Percent Range 90%
Packaging Amber Glass Bottle
Linear Formula (CH3)2CHC(=NH)NH2·HCl
Quantity 1 g
Formula Weight 122.6
Percent Purity 90%
Chemical Name or Material 2-Methylpropanimidamide hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.