missing translation for 'onlineSavingsMsg'
Learn More

(2-Methylpropyl)boronic acid, 95+%

Product Code. 10364592 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10364592 1 g Glass bottle 1g
10467782 5 g Glass bottle 5g
10370474 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10364592 Supplier Thermo Scientific Supplier No. 359950010
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 84110-40-7 | C4H11BO2 | 101.94 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 84110-40-7
Molecular Formula C4H11BO2
Molecular Weight (g/mol) 101.94
MDL Number MFCD00134156
InChI Key ZAZPDOYUCVFPOI-UHFFFAOYSA-N
Synonym isobutylboronic acid, isobutaneboronic acid, 2-methylpropyl boronic acid, 1-isobutaneboronic acid, 2-methylpropaneboronic acid, 2-methylprop-1-ylboronic acid, unii-khx76h7jhz, khx76h7jhz, 2-methyl-1-propylboronic acid, 2-methyl-1-propaneboronic acid
PubChem CID 2734395
IUPAC Name 2-methylpropylboronic acid
SMILES CC(C)CB(O)O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 108°C to 111°C
Color White to Cream
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 1 g
Beilstein 04,III,1965
Solubility Information Solubility in water: soluble. Other solubilities: soluble in methanol
Formula Weight 101.94
Percent Purity 98%
Physical Form Crystalline Powder or Flakes
Chemical Name or Material (2-Methylpropyl)boronic acid
Show More Show Less
compliance-icons
Product Identifier
  • (2-Methylpropyl)boronic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.