missing translation for 'onlineSavingsMsg'
Learn More

2-Methylresorcinol, 98%

Product Code. 10723023 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10723023 100 g Glass bottle 100g
10242422 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10723023 Supplier Thermo Scientific Acros Supplier No. 364741000
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 608-25-3 | C7H8O2 | 124.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 608-25-3
Molecular Formula C7H8O2
Molecular Weight (g/mol) 124.14
MDL Number MFCD00002271
InChI Key ZTMADXFOCUXMJE-UHFFFAOYSA-N
Synonym 2-methylresorcinol, 2,6-dihydroxytoluene, 1,3-benzenediol, 2-methyl, 2-methylresorcin, resorcinol, 2-methyl, toluene-2,6-diol, 2-methyl-1,3-benzenediol, 1,3-dihydroxy-2-methylbenzene, 1,3-benzenediol, methyl, 2-methyl resorcinol
PubChem CID 11843
IUPAC Name 2-methylbenzene-1,3-diol
SMILES CC1=C(C=CC=C1O)O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 118°C to 124°C
Color White
Boiling Point 264°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3C6H3(OH)2
Quantity 100 g
Beilstein 06, 878
Formula Weight 124.14
Percent Purity 98%
Physical Form Glassy Irregular Shaped Granules
Chemical Name or Material 2-Methylresorcinol, 98%
Show More Show Less
compliance-icons
Product Identifier
  • 2-Methylresorcinol
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.