missing translation for 'onlineSavingsMsg'
Learn More

2-Methylresorcinol, 98%

Product Code. 11449413
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
Unit Size:
1000g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11449413 250 g 250g
11439413 50 g 50g
11429413 1000 g 1000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11449413 Supplier Thermo Scientific Alfa Aesar Supplier No. A11821.30
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 608-25-3 | C7H8O2 | 124.139 g/mol

2-Methylresorcinol is used in the preparation of aromatic benziporphyrins, tripyrrane analogs and C-5-bromo-2-hydroxyphenylcalix[4]-2-methylresorcinarene. Further, it is employed in oxidative and non-oxidative hair dye formulations.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Methylresorcinol is used in the preparation of aromatic benziporphyrins, tripyrrane analogs and C-5-bromo-2-hydroxyphenylcalix[4]-2-methylresorcinarene. Further, it is employed in oxidative and non-oxidative hair dye formulations.

Solubility
Soluble in water and methanol.

Notes
Incompatible with acid chlorides, acid anhydrides and oxidizing agents
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 608-25-3
Molecular Formula C7H8O2
Molecular Weight (g/mol) 124.139
MDL Number MFCD00002271
InChI Key ZTMADXFOCUXMJE-UHFFFAOYSA-N
Synonym 2-methylresorcinol, 2,6-dihydroxytoluene, 1,3-benzenediol, 2-methyl, 2-methylresorcin, resorcinol, 2-methyl, toluene-2,6-diol, 2-methyl-1,3-benzenediol, 1,3-dihydroxy-2-methylbenzene, 1,3-benzenediol, methyl, 2-methyl resorcinol
PubChem CID 11843
IUPAC Name 2-methylbenzene-1,3-diol
SMILES CC1=C(C=CC=C1O)O

Specifications

Melting Point 118°C to 122°C
Boiling Point 264°C
Flash Point 135°C (275°F)
Quantity 250 g
UN Number UN2811
Beilstein 2042177
Solubility Information Soluble in water and methanol.
Formula Weight 124.14
Percent Purity 98%
Chemical Name or Material 2-Methylresorcinol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.