missing translation for 'onlineSavingsMsg'
Learn More

2-Methylsulphonylaniline hydrochloride, 95%

Product Code. 10118240 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
2.5 g
Packaging:
Glass bottle
Unit Size:
2.5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10118240 2.5 g Glass bottle 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10118240 Supplier Thermo Scientific Acros Supplier No. 424840025
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 205985-95-1 | C7H9NO2S | 171.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 205985-95-1
CAS Min % 94.0
CAS Max % 100.0
Melting Point 139°C to 141°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 94% min. (HPLC)
Packaging Glass bottle
Molecular Formula C7H9NO2S
Linear Formula SO2CH3C6H4NH2·HCl
MDL Number MFCD00216484
Quantity 2.5 g
Synonym 2-methylsulfonylaniline hydrochloride, 2-methylsulfonyl aniline hydrochloride, 2-methanesulfonylaniline hydrochloride, 2-methylsulphonylaniline hydrochloride, 2-methylsulphonyl aniline hydrochloride, 2-methylsulfonyl aniline, hcl, 2-methanesulfonyl-phenylammonium chloride, 2-methylsulfonyl phenylamine hydrochloride
Solubility Information Solubility in water: slight
InChI Key AUSRSVRJOXYXMI-UHFFFAOYSA-N
SMILES CS(=O)(=O)C1=CC=CC=C1N
IUPAC Name 2-methylsulfonylaniline;hydrochloride
Molecular Weight (g/mol) 171.21
PubChem CID 2735177
Formula Weight 207.68
Percent Purity 95%
Physical Form Powder
Chemical Name or Material 2-Methylsulphonylaniline hydrochloride, 95%
Show More Show Less
compliance-icons
Product Identifier
  • 2-Methylsulphonylaniline hydrochloride
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.