missing translation for 'onlineSavingsMsg'
Learn More

2-Methyltetrahydrofuran-3-one, 98+%

Product Code. 11439926
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11439926 5 g 5g
11449926 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11439926 Supplier Thermo Scientific Alfa Aesar Supplier No. B20890.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 3188-00-9 | C5H8O2 | 100.117 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3188-00-9
Molecular Formula C5H8O2
Molecular Weight (g/mol) 100.117
MDL Number MFCD00010423
InChI Key FCWYQRVIQDNGBI-UHFFFAOYSA-N
Synonym 2-methyltetrahydrofuran-3-one, 2-methyl tetrahydro-3-furanone, 2-methyltetrahydro-3-furanone, 3 2h-furanone, dihydro-2-methyl, dihydro-2-methyl-3 2h-furanone, coffee furanone, dihydro-2-methyl-3-furanone, tetrahydro-2-methylfuran-3-one, 2-methyl-3-oxotetrahydrofuran, 2-methyl-3-ketotetrahydrofuran
PubChem CID 18522
IUPAC Name 2-methyloxolan-3-one
SMILES CC1C(=O)CCO1

Specifications

Density 1.034
Quantity 5 g
Odor Characteristic
Refractive Index 1.429
UN Number UN1224
Beilstein 1341334
Boiling Point 138°C to 139°C
Formula Weight 100.12
Percent Purity ≥98%
Chemical Name or Material 2-Methyltetrahydrofuran-3-one

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.